Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Storage Boxes

Red, 600×400×190 mm
Catalog # CORAG119709
Supplier:   Gilac PROFESSIONNEL
Storage Boxes
Catalog # CORAG119709
Storage Boxes
Storage Boxes

Red, 600×400×190 mm

Price
109,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.

  • Very good chemical resistance
  • Very good thermal resistance
  • Very sturdy and economical
Specifications
  • Colour:
    Red
  • Seq:
    09
  • W×D×H:
    600×400×190 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.