Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Green, 395×295×160 mm
Catalog # 216-2869
Supplier:   Gilac PROFESSIONNEL
Storage Boxes
Catalog # 216-2869
Storage Boxes
Storage Boxes

Green, 395×295×160 mm

Price
54,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.

  • Very good chemical resistance
  • Very good thermal resistance
  • Very sturdy and economical
Specifications
  • Colour:
    Green
  • Seq:
    03
  • W×D×H:
    395×295×160 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.