ContactUsLinkComponent
Sensolyt® pt,orp combination electrode for heavily polluted wastewater, for use in flow-thru vessels: d 700/n, d 530, d 300/ph etc.w ith watertight s7 plug head (ip67) 1 * 1 items
Catalog # WTWO105412
Supplier:  WTW ONLINE
Sensolyt® pt,orp combination electrode for heavily polluted wastewater, for use in flow-thru vessels: d 700/n, d 530, d 300/ph etc.w ith watertight s7 plug head (ip67) 1 * 1 items
Catalog # WTWO105412
Supplier:  WTW ONLINE
Frequently Bought Together