Order Entry
Austria
Orders LinkContactUsLinkComponent
Handheld refractometers, Eclipse

Handheld refractometers, Eclipse

Handheld refractometer for starch, 0 − 30 %, 0,2 %
Catalog # 635-0048
Supplier:   BELLINGHAM STANLEY
Handheld refractometers, Eclipse
Catalog # 635-0048
Supplier:  BELLINGHAM STANLEY
Handheld refractometers, Eclipse
Handheld refractometers, Eclipse

Handheld refractometer for starch, 0 − 30 %, 0,2 %

Price
294,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Handheld refractometers use high precision clear scale, and simple zero adjustment with lock.

  • Shaded field of view for distinction
  • Anti-roll support and ergonomic design
  • Comfortable eyepiece with smooth adjustment
  • Easy to clean prism and push-on flap protection
  • Unique sample ‘dribble’ feature

Delivery information: Supplied with a soft carrying case and instructions.

Specifications
  • Range:
    0 − 30 %
  • Resolution:
    0,2 %
  • Description:
    Handheld refractometer for starch
  • Seq:
    10