Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 44, excitation BP 475/40, beam splitter FT 500, emission BP 530/50
Catalog # ZEIS0000001114459
Supplier:   ZEISS
Filter sets
Catalog # ZEIS0000001114459
Supplier:  ZEISS
Filter sets
Filter set 44, excitation BP 475/40, beam splitter FT 500, emission BP 530/50
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Filter set 44, excitation BP 475/40, beam splitter FT 500, emission BP 530/50
  • Seq:
    11