Order Entry
Austria
Orders LinkContactUsLinkComponent
Cooling systems, CoolSafe™

Cooling systems, CoolSafe™

Cool brick -10 °C, 0,2/0,5/1,5 and 5 ml tubes
Catalog # DIVEBRIK-1520
Supplier:   DIVERSIFIED BIOTECH
Cooling systems, CoolSafe™
Catalog # DIVEBRIK-1520
Cooling systems, CoolSafe™
Cooling systems, CoolSafe™

Cool brick -10 °C, 0,2/0,5/1,5 and 5 ml tubes

Price
121,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Polystyrene cool safe box, aluminium cooling chamber.

  • Durable
  • Temperature stable
  • Corrosion resistant

Cooling system designed for 0,2 ml tubes (strips), 96 and 384 well microplates, 0,5, 1,5 and 5,0 ml tubes. Cool bricks with different temperature zones 1 or −10 °C.

Specifications
  • For:
    0,2/0,5/1,5 and 5 ml tubes
  • Description:
    Cool brick -10 °C
  • Seq:
    13
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.