Draining racks
Draining racks, for 60 test tubes and 5 flasks, 420×170×610 mm
Catalog # HECH49027010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Made of wire, plastic coated.
- Good chemical resistance
- Good temperature resistance
- Easy to clean
Suitable for hanging or free standing test tubes and flasks with a plastic trough.
Specifications
- Description:Draining racks, for 60 test tubes and 5 flasks
- Seq:01
- W×D×H:420×170×610 mm