Order Entry
Austria
Orders LinkContactUsLinkComponent
Staining plates

Staining plates

12, Ø×D: 15×0,8 mm, 89×57×4 mm
Catalog # HECH40624010
Staining plates
Catalog # HECH40624010
Staining plates
Staining plates

12, Ø×D: 15×0,8 mm, 89×57×4 mm

Price
31,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Glass spot plates with 10 or 12 numbered cavities. For flocculation tests.

Specifications
  • No. of wells:
    12
  • Seq:
    02
  • W×D×H:
    89×57×4 mm
  • Well size:
    Ø×D: 15×0,8 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.