Order Entry
Austria
Orders LinkContactUsLinkComponent
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®

Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®

Glass bottle, Safebreak®, 2,5 L
Catalog # 20910.328
Health
Toxic
Corrosive
Supplier:   VWR Chemicals
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Catalog # 20910.328
Supplier:  VWR Chemicals
CAS Number:  50-00-0
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®

Glass bottle, Safebreak®, 2,5 L

CAS Number:  50-00-0
Price
54,10 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Density:1.088 g/cm³ (20 °C)
  • Flash Pt:66 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
  •  :Stabilised with methanol 10 %
Specifications

Specification Test Results

  • Assay:
    35 - 38 %
  • Density (20/4):
    1.083 - 1.093
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.02 %
  • Methanol:
    8 - 12 %

Specifications

  • Pack type:
    Glass bottle, Safebreak®
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.