Order Entry
Austria
Orders LinkContactUsLinkComponent
pH buffer solutions, NIST traceable, Orion™

pH buffer solutions, NIST traceable, Orion™

pH 4,01 buffer, colour-coded pink (5×60 ml bottle), 4,01, Pink, Plastic bottle, 5
Artikelnr. ORIO910460
Anbieter:   Thermo Orion
pH buffer solutions, NIST traceable, Orion™
Artikelnr. ORIO910460
Anbieter:  Thermo Orion
CAS Nummer:  
pH buffer solutions, NIST traceable, Orion™
pH buffer solutions, NIST traceable, Orion™

pH 4,01 buffer, colour-coded pink (5×60 ml bottle), 4,01, Pink, Plastic bottle, 5

CAS Nummer:  
Preis
55,40 €
Log in for availability
1 * 5 ST
Product Details & Documents

Perform accurate, reproducible pH calibrations with Orion™ pH buffers.

  • NIST traceability
  • Colour-coded solutions are available for ease of use and identification

Solutions are manufactured under ISO 9000 quality standards.

The kit includes 1 pint (475 ml) bottle each of pH 4,00 buffer, pH 7,00 buffer, pH 10,01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.

Delivery information: Supplied in 475 ml or 60 ml bottles, with buffer/temperature tables to ensure more precise calibration.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5
  • Colour:
    Pink
  • Description:
    pH 4,01 buffer, colour-coded pink (5×60 ml bottle)
  • pH value:
    4,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.