Orders LinkContactUsLinkComponent
Pipette Tips, Traditional
Pipette Tips, Traditional
Standard, 10 ml, -, RAININ®, Pipetman® and other conical shaft pipettes, 10 ml, Bulk, 200/bag
Catalog # 613-4749
Marketsource
Supplier:   RAININ
Pipette Tips, Traditional
Catalog # 613-4749
Supplier:  RAININ
Marketsource
Pipette Tips, Traditional
Standard, 10 ml, -, RAININ®, Pipetman® and other conical shaft pipettes, 10 ml, Bulk, 200/bag
Price
Inquire For Price
Log in for availability
1 * 200 ST
Product Details & Documents
These thin-wall tips are designed to fit RAININ and Pipetman pipettes with conical shafts, Eppendorf and Labsystems pipettes. Some tips feature a hydrophobic filter which forms a barrier against aerosols and liquids eliminating potential sample and pipette cross contamination.
  • Filtered and pre-sterilised bioclean tips are gamma-irradiated
  • ShaftGard tips are longer than standard tips for easy access into microcentrifuge and PCR tubes
  • Wide orifice tips minimise shear forces on delicate samples and flow resistance for viscous samples
The tips are available in different packaging options:
Racks with removable cover
- Sealed bottoms and overlapping lids protect tips from dust, dirt and micro-organisms
- Two-position lid allows autoclaving with the lid in vent position
- Sturdy shelf construction allows easy tip loading with single and multi-channel pipettes
Green-Pak® Individual Refills and Green-Pak® SpaceSaver® Refills
- Environmentally friendly refills for racks; less plastic waste
- Individual refills contain 96 tips in 8×12 format, sealed in a shell of Code 1 PET recyclable plastic
- Each refill is completely sealed to protect the tips; hands and fingers never come in contact with tips during loading
- SpaceSaver® systems are provided with either eight or ten stacked refills
- SpaceSaver® refills are packaged in a code 1 PET sleeve which functions as an automatic tip dispenser that aligns and refills an empty rack with 96 tips at a time
Bulk
- Packaged in zip-lock bags
- Very economical
Specifications
  • Packed:
    Bulk, 200/bag
  • Sterile:
    -
  • Volume:
    10 ml
  • Seq:
    102
  • Tip style:
    Standard
  • For use with:
    RAININ®, Pipetman® and other conical shaft pipettes, 10 ml