Orders LinkContactUsLinkComponent
Test kits, digital titration method
Test kits, digital titration method
Chloride testing: 1,5-diphenyl carbazone powder pillows by mercuric nitrate titration method, 10 - 8000  mg/l, 100
Catalog # 705-0240
Supplier:   Hach
Test kits, digital titration method
Catalog # 705-0240
Supplier:  Hach
CAS Number:  
Test kits, digital titration method
Test kits, digital titration method
Chloride testing: 1,5-diphenyl carbazone powder pillows by mercuric nitrate titration method, 10 - 8000  mg/l, 100
CAS Number:  
Price
92,50 €
Log in for availability
1 * 100 ST
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    100
  • Description:
    Chloride testing: 1,5-diphenyl carbazone powder pillows by mercuric nitrate titration method
  • Range:
    10 - 8000  mg/l