Test kits, digital titration method
Sulphite: iodate-iodide 0.3998 N cartridge, 1
Catalog # 705-0074
Supplier: Hach
Jump to:
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of colour
- Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
- Pk:1
- Description:Sulphite: iodate-iodide 0.3998 N cartridge