Orders LinkContactUsLinkComponent
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
15-415, Purple
Catalog # 215-4017
Supplier:   DWK Life Sciences
Dropper tip caps, WHEATON®
Catalog # 215-4017
Supplier:  DWK Life Sciences
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
15-415, Purple
Price
51,20 €
Log in for availability
1 * 100 ST
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specifications
  • Thread:
    15-415
  • Seq:
    30
  • Cap colour:
    Purple