Orders LinkContactUsLinkComponent
Lifting platforms for Lauda baths
Lifting platforms for Lauda baths
Lifting platform, for 1× E 106 T/E 206 T/ET 6 and 2× A 24, 110×260 mm
Catalog # 461-2951
Supplier:   LAUDA
Lifting platforms for Lauda baths
Catalog # 461-2951
Supplier:  LAUDA
Lifting platforms for Lauda baths
Lifting platforms for Lauda baths
Lifting platform, for 1× E 106 T/E 206 T/ET 6 and 2× A 24, 110×260 mm
Price
414,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These platforms are made from stainless steel. For use with Lauda baths.
  • Height adjustable in eight steps
  • To be fixed on one side of the bath
  • Not for use with bath cover or gable cover
Specifications
  • Description:
    Lifting platform, for 1× E 106 T/E 206 T/ET 6 and 2× A 24
  • L×W:
    110×260 mm
  • Seq:
    02