Orders LinkContactUsLinkComponent
Lifting platforms for Lauda baths
Lifting platforms for Lauda baths
Lifting platform, for 1× E 4/E 103/E 203; 2× A 12 and 6× A 24, 115×70 mm
Catalog # 461-2948
Supplier:   LAUDA
Lifting platforms for Lauda baths
Catalog # 461-2948
Supplier:  LAUDA
Lifting platforms for Lauda baths
Lifting platforms for Lauda baths
Lifting platform, for 1× E 4/E 103/E 203; 2× A 12 and 6× A 24, 115×70 mm
Price
406,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These platforms are made from stainless steel. For use with Lauda baths.
  • Height adjustable in eight steps
  • To be fixed on one side of the bath
  • Not for use with bath cover or gable cover
Specifications
  • Description:
    Lifting platform, for 1× E 4/E 103/E 203; 2× A 12 and 6× A 24
  • L×W:
    115×70 mm
  • Seq:
    01