Order Entry
Austria
Orders LinkContactUsLinkComponent
Manual cell counters

Manual cell counters

Manual cell counter, 5
Catalog # 720-0164
Supplier:   MOINEAU
Manual cell counters
Catalog # 720-0164
Supplier:  MOINEAU
Manual cell counters
Manual cell counters

Manual cell counter, 5

Price
449,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This manual cell counter can be used in combination of up to five counters.

  • Four digit display (4,3×3,5 mm) with reset knob
  • Use finger pressure
  • Can be fixed in position

W×D×H: 37,6×0,4×38,1 mm

Specifications
  • Description:
    Manual cell counter
  • Array:
    5