Flasks, flat bottom with Quickfit® ground joint
250 ml, 120 mm, 24/29 NS
Catalog # 201-8443
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, clear, with short neck.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat base allows use with hotplates. Labelling area.
Specifications
- Socket size:24/29 NS
- Capacity:250 ml
- Height:120 mm
- Seq:12



