Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Wire baskets

240×180 mm
  216-2845
 :   BOCHEM
Wire baskets
  216-2845
 :  BOCHEM
Wire baskets
Wire baskets

240×180 mm

 
129,00 €
   
1 * 1 ST
 

Stainless steel.

  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Stowable handle

These round baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.

 
  • Ø×H:
    240×180 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.