Wire baskets
150×120 mm
Catalog # 216-2842
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel.
- Spot-welded grid 8×8×1 mm
- Reinforced edges
- Stowable handle
These round baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
Specifications
- Ø×H:150×120 mm
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



