Order Entry
Austria
Orders LinkContactUsLinkComponent
Wire baskets

Wire baskets

100×80 mm
Catalog # 216-2840
Supplier:   BOCHEM
Wire baskets
Catalog # 216-2840
Supplier:  BOCHEM
Wire baskets
Wire baskets

100×80 mm

Price
57,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel.

  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Stowable handle

These round baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.

Specifications
  • Ø×H:
    100×80 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.