Orders LinkContactUsLinkComponent
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
8-425, White
Catalog # 215-4011
Supplier:   DWK Life Sciences
Dropper tip caps, WHEATON®
Catalog # 215-4011
Supplier:  DWK Life Sciences
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
8-425, White
Price
48,30 €
Log in for availability
1 * 100 ST
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specifications
  • Thread:
    8-425
  • Seq:
    09
  • Cap colour:
    White