Order Entry
Austria
Orders LinkContactUsLinkComponent
Dropping bottles, LDPE, WHEATON®

Dropping bottles, LDPE, WHEATON®

60 ml, LDPE, translucent, 20-410
Catalog # 215-8002
Supplier:   DWK Life Sciences
Dropping bottles, LDPE, WHEATON®
Catalog # 215-8002
Supplier:  DWK Life Sciences
Dropping bottles, LDPE, WHEATON®
Dropping bottles, LDPE, WHEATON®

60 ml, LDPE, translucent, 20-410

Price
591,00 €
Log in for availability
1 * 144 ST
Product Details & Documents

LDPE, translucent, with white PP cap.

  • Good chemical resistance, heat resistant up to 80 °C
  • Bottle dispenses one drop at a time
  • Dropper tip snaps firmly in place and is easily removed for filling

With extended controlled dropper tip and dropper tip cap. These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications.

Documents

Specifications
  • Thread:
    20-410
  • Capacity:
    60 ml
  • Material:
    LDPE, translucent
  • Seq:
    09
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.