Orders LinkContactUsLinkComponent
Charts
Charts
C652, –50...+50 °C, 24 hours
Catalog # 620-1542
Supplier:   DICKSON
Charts
Catalog # 620-1542
Supplier:  DICKSON
Charts
C652, –50...+50 °C, 24 hours
Price
79,10 €
Log in for availability
1 * 60 ST
Product Details & Documents
Specifications
  • Description:
    C652
  • Seq:
    02
  • Range:
    –50...+50 °C
  • Recording time:
    24 hours