0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
Orders Link
ContactUsLinkComponent
Closures
Bottle Closures
Screw caps, bakelite
Screw caps, bakelite
with Butyl/PTFE seal, 35 mm, Black, Bakelite
215-2684
:
GRAVIS
Screw caps, bakelite
215-2684
:
GRAVIS
Screw caps, bakelite
with Butyl/PTFE seal, 35 mm, Black, Bakelite
306,00 €
1 * 100 ST
-
+
SPVAP08028_TDS_EN
Thread:
35 mm
Colour:
Black
Description:
with Butyl/PTFE seal
Material:
Bakelite
Seq:
06
Bottles, round with wide neck or flat with narrow neck
#: SPVAB00354EURO
: GRAVIS