Order Entry
Austria
Orders LinkContactUsLinkComponent
Sample dippers

Sample dippers

Dip sampler for liquid samples, Transparent, PP, 1000 mm, 22 mm
Catalog # BURK5315-0100
Supplier:   BURKLE
Sample dippers
Catalog # BURK5315-0100
Supplier:  BURKLE
Sample dippers
Sample dippers

Dip sampler for liquid samples, Transparent, PP, 1000 mm, 22 mm

Price
59,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

Plunging siphons for the simple removal of liquid samples.

  • Made from transparent PP
  • All-layer and point sampling
  • Upper end with reflux valve, lower end with ball valve
Specifications
  • Ø:
    22 mm
  • Colour:
    Transparent
  • Description:
    Dip sampler for liquid samples
  • Length:
    1000 mm
  • Material:
    PP
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.