ContactUsLinkComponent
Vertical Dual adjustable slab gel kit 14,5-40cm. Supplements included: 1x DASG-CLAMPS: Dual Slab Gel, 4ea #1, 12ea #2 1x INSTRUCTIONS: Kit/Unit Instructions 2x NGP-400NR: GW/notchd plate st 16.5x38.7cm 1x RODS, 400: Guide rods for ASG/DASG, 400mm 2x 1 * 1 items
Catalog # 730-0007
Supplier:  Expedeon
Vertical Dual adjustable slab gel kit 14,5-40cm. Supplements included: 1x DASG-CLAMPS: Dual Slab Gel, 4ea #1, 12ea #2 1x INSTRUCTIONS: Kit/Unit Instructions 2x NGP-400NR: GW/notchd plate st 16.5x38.7cm 1x RODS, 400: Guide rods for ASG/DASG, 400mm 2x 1 * 1 items
Catalog # 730-0007
Supplier:  Expedeon
Frequently Bought Together