Order Entry
Austria
Orders LinkContactUsLinkComponent
Drying tubes, U-shaped

Drying tubes, U-shaped

130, 13 mm
Catalog # LENZ05339013
Supplier:   Lenz Laborglas
Drying tubes, U-shaped
Catalog # LENZ05339013
Supplier:  Lenz Laborglas
Drying tubes, U-shaped
Drying tubes, U-shaped

130, 13 mm

Price
24,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Drying tubes, U-shaped, side tubes with olives (Ext.Ø: 6 mm).

Documents

Specifications
  • Ø int.:
    13 mm
  • Length:
    130
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.