Orders LinkContactUsLinkComponent
Packaging and Storage Boxes
Packaging and Storage Boxes
Transparent, 94×68×29 mm, PS
Catalog # SEMA1729
Supplier:   SEMADENI
Packaging and Storage Boxes
Catalog # SEMA1729
Supplier:  SEMADENI
Packaging and Storage Boxes
Packaging and Storage Boxes
Transparent, 94×68×29 mm, PS
Price
2,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
These boxes made of polystyrene (PS) are crystal clear with a good closing tight fitting lid.
  • Clear for easy product identification
  • The lids are either mounted or not mounted
  • Stackable
Specifications
  • Colour:
    Transparent
  • Material:
    PS
  • Seq:
    14
  • W×D×H:
    94×68×29 mm