Dispersing elements (aggregates) for POLYTRON® PT models
PT-DA 12/2WEC-E157, PT 1200 E / PT 2500 E, 14,1 m/s, 12/9 mm, 154 mm, 2-250 ml
431-0057
Designation: EC - EasyClean version for the simplest cleaning or sterilisation (Standard)/2 - rotor/stator with 1 tooth row each.
: Other dispersing aggregates available on request.
- Speed:14,1 m/s
- Volume:2-250 ml
- Description:PT-DA 12/2WEC-E157
- Length:154 mm
- Seq:08
- Stator/rotor Ø:12/9 mm
- For unit:PT 1200 E / PT 2500 E
- Cat. No.:431-0057
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



