Orders LinkContactUsLinkComponent
Syringes, SampleLock™
Syringes, SampleLock™
22, 10000 µl, 51 mm, 2, 1010 RN
  HAMI81656
Syringes, SampleLock™
  HAMI81656
Syringes, SampleLock™
Syringes, SampleLock™
22, 10000 µl, 51 mm, 2, 1010 RN
394,00 €
1 * 1 ST
These syringes allow the collection, storage, transport and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
  • Not autoclavable, gas sterilisable
  • An easy-to-use twist valve
  • A positive rear stop prevents loss of sample and plunger blowout
  • Fluid path includes SST, borosilicate glass and PTFE
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking.
Optional male luer and female luer lock adapters (sold separately) threads onto the SampleLock™ valve, making the syringes compatible with a multitude of connectors and fittings.
  • Gauge:
    22
  • Type:
    1010 RN
  • Type of tip:
    2
  • Volume:
    10000 µl
  • Length:
    51 mm
  • Seq:
    04
Accessories
  •  
    Description
  • HAMI35081
    Removable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock termination
    1 * 1 ST - 56,70 €
  •  
    Description
  • HAMI35080
    Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Tip (LT) termination
    1 * 1 ST - 46,40 €
  •  
    Description
  • HAMI35083
    Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Lock (TLL) termination
    1 * 1 ST - 50,50 €
  •  
    Gauge
    Length
    Ext.Ø
  • HAMI7784-02
    22
    51 mm
    0,72 mm
    1 * 6 ST - 230,00 €
  •  
    Gauge
    Length
    Ext.Ø
  • HAMI7779-01
    22
    51 mm
    0,72 mm
    1 * 6 ST - 79,40 €
  •  
    Gauge
    Length
    Ext.Ø
  • HAMI7784-01
    22s
    51 mm
    0,72 mm
    1 * 6 ST - 234,00 €
  •  
    Gauge
    Length
    Ext.Ø
  • HAMI7779-03
    22s
    51 mm
    0,72 mm
    1 * 6 ST - 79,40 €
  •  
    Gauge
    Length
    Ext.Ø
  • HAMI7780-03
    22s
    51 mm
    0,72 mm
    1 * 6 ST - 62,90 €
  •  
    Gauge
    Length
    Ext.Ø
  • HAMI7780-04
    22
    51 mm
    0,72 mm
    1 * 6 ST - 60,80 €