ContactUsLinkComponent
Ess 700 va/n,stainless steel (1.4571) weld-in socket, for measurements in pipelines, for use with all sensors of 650/690/700(iq)/701 (iq) seriesplease order suitable sensor flange separately 1 * 1 items
Catalog # WTWO203755
Supplier:  WTW ONLINE
Ess 700 va/n,stainless steel (1.4571) weld-in socket, for measurements in pipelines, for use with all sensors of 650/690/700(iq)/701 (iq) seriesplease order suitable sensor flange separately 1 * 1 items
Catalog # WTWO203755
Supplier:  WTW ONLINE
Frequently Bought Together