ContactUsLinkComponent
Compound eye, t.s. through head of worker (Apis mellifica), showing the structure of the typical insect eyes and brain. Ommatidia ar e seen. 1 * 1 items
Catalog # LIEDIN251E
Supplier:  LIEDER JOHANNES
Compound eye, t.s. through head of worker (Apis mellifica), showing the structure of the typical insect eyes and brain. Ommatidia ar e seen. 1 * 1 items
Catalog # LIEDIN251E
Supplier:  LIEDER JOHANNES
Frequently Bought Together