Order Entry
Austria
Orders LinkContactUsLinkComponent
ORP electrode solutions

ORP electrode solutions

ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 470 mV, 250 mL
Catalog # SCOI285138168
Supplier:   SI Analytics
ORP electrode solutions
Catalog # SCOI285138168
Supplier:  SI Analytics
CAS Number:  
ORP electrode solutions
ORP electrode solutions

ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 470 mV, 250 mL

CAS Number:  
Price
54,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

ORP electrode solutions are used as cleaning solutions for combination electrodes and reference electrodes.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 470 mV

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.