Orders LinkContactUsLinkComponent
Colorimetric test kit, sulphite, TESTOVAL®
Colorimetric test kit, sulphite, TESTOVAL®
Sulphite, refill pack, 0,5-1-2,5-5-10-20 mg/l, 150 Tests
Catalog # HEYL410635
Colorimetric test kit, sulphite, TESTOVAL®
Catalog # HEYL410635
CAS Number:  
Colorimetric test kit, sulphite, TESTOVAL®
Colorimetric test kit, sulphite, TESTOVAL®
Sulphite, refill pack, 0,5-1-2,5-5-10-20 mg/l, 150 Tests
CAS Number:  
Price
97,60 €
Log in for availability
1 * 150 Tests
Product Details & Documents
Visual colour test method using a comparator disc. Two reagents.
Specifications
Specifications
  • Pk:
    150 Tests
  • Description:
    Sulphite, refill pack
  • Range:
    0,5-1-2,5-5-10-20 mg/l
Specifications
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Replacement cuvettes for colour comparison kits
    Replacement cuvettes for colour comparison kits
    HEYL410001
    1
    Log in for availability
    1 * 1 ST - 12,70 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Replacement colour comparator, complete
    Replacement colour comparator, complete
    HEYL410636
    1
    Log in for availability
    1 * 1 ST - 77,10 €