Order Entry
Austria
Orders LinkContactUsLinkComponent
Celite® 545, filter aid

Celite® 545, filter aid

Glass bottle for solids, 2,5 kg
Catalog # ROTH0011.1
Health
Marketsource
Supplier:   Roth Carl
Celite® 545, filter aid
Catalog # ROTH0011.1
Supplier:  Roth Carl
Marketsource
CAS Number:  68855-54-9
Celite® 545, filter aid

Glass bottle for solids, 2,5 kg

CAS Number:  68855-54-9
Price
47,70 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  DE, Diatomaceous earth, flux-calcined
  • CAS Number:68855-54-9
  • Formula:SiO₂
  • MDL Number:MFCD00132803
  • Molecular Weight:60.08 g/mol
  • EINECS:272-489-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.