INDICATOR 500ML TYPE 301 TESTOMAT 1 * 500 mL
713-1302
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Accessories
: Hach