BUFFER SOLUTION REDOX UH=427MV PH=10 1 * 250 mL
Catalog # 662-8152
Supplier: Mettler - Toledo
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Redox buffer solutions
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
ORP BUFFER 271 MV, 500 ML 1 * 500 mL
Supplier: HAMILTON BONADUZ