[DE]PH-Pufferlösung 4.60, 250ML 1 * 250 ml
Catalog # 662-8123
Supplier: Mettler - Toledo
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Hypodermic needles, HENKE-JECT® for single use
Supplier: HENKE SASS WOLF
Dehydrated culture media
Supplier: Merck
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH buffers, ready to use
Supplier: WTW