Rectangular cover glasses, Menzel Gläser
Cover glasses, 24×55 mm, #1
Catalog # 631-9444
Supplier: Epredia
Jump to:
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
- Uniform quality of thickness
- No bubbles or flaws
- Made of pre-washed pure white Hydrolytic Class 1 glass
Documents
Specifications
- No.:#1
- Description:Cover glasses
- L×W:24×55 mm