Rectangular cover glasses, Menzel Gläser
Cover glasses, 24×50 mm, #1,5
631-9430
: Epredia
These cover slips are suitable for in vitro diagnostic (IVD) applications.
- Uniform quality of thickness
- No bubbles or flaws
- Made of pre-washed pure white Hydrolytic Class 1 glass
- No.:#1,5
- Description:Cover glasses
- L×W:24×50 mm
- Seq:29