Orders LinkContactUsLinkComponent
Proline® Single Channel Pipettes, Mechanical, Fixed/Variable Volume
500 μl, 99,7%, 0,2%, 1000 / F1000 µl, 613-4069, 613-4074
  613-4086
   Low Manufacturing Impact
 :   Sartorius
Proline® Single Channel Pipettes, Mechanical, Fixed/Variable Volume
  613-4086
 :  Sartorius
   Low Manufacturing Impact
Proline® Single Channel Pipettes, Mechanical, Fixed/Variable Volume
Proline® Single Channel Pipettes, Mechanical, Fixed/Variable Volume
500 μl, 99,7%, 0,2%, 1000 / F1000 µl, 613-4069, 613-4074
183,00 €
1 * 1 ST
Ergonomic pipettes with fast, easy operation. Selected models offer changeable tip cone filters to prevent liquids and aerosols entering the pipette.
  • Easy calibration and maintenance, small number of parts
  • Smooth plunger action and simple in-lab recalibration and service
  • Volume setting by a simple click-stop
  • Ergonomic finger support, which reduces the risk of RSI
Two types of filters are offered: Safe-Cone standard filter and Safe-Cone Plus filter. It is recommended to use the Standard filter for general applications and the Plus filter for more demanding applications, such as cell culture, bacterial, viral work and molecular biology.
 : Supplied with five extra filters, forceps and a calibration tool.
  • Volume:
    500 μl
  • Seq:
    18
  • Accuracy:
    99,7%
  • Imprecision:
    0,2%
  • Recommended tips:
    1000 / F1000 µl
  • With Safe-Cone filter(s):
    613-4069, 613-4074
  • Cat. No.:
    613-4086
  • Environmental certification:
    Renewable energy certificate; ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
  •  
    Description
  • 613-4068
    Safe-Cone filters, Standard, Ø 6,73 mm
    1 * 50 ST - 37,10 €
  •  
    Description
  • 613-4070
    Safe-Cone filters, Standard, Ø 3,15 mm
    1 * 50 ST - 37,10 €
  •  
    Description
  • 613-0694
    Safe-Cone filters, Plus, Ø 2,51 mm
    1 * 50 ST - 45,60 €
  •  
    Description
  • 613-4071
    Safe-Cone filters, Standard, Ø 2,51 mm
    1 * 50 ST - 37,10 €
  •  
    Description
  • 613-4069
    Safe-Cone filters, Standard, Ø 5,33 mm
    1 * 50 ST - 37,10 €
  •  
    Description
  • 613-0692
    Safe-Cone filters, Plus, Ø 6,73 mm
    1 * 50 ST - 45,60 €
  •  
    Description
  • 613-4074
    Safe-Cone filters, Plus, Ø 5,33 mm
    1 * 50 ST - 45,60 €
  •  
    Description
  • 613-0693
    Safe-Cone filters, Plus, Ø 3,15 mm
    1 * 50 ST - 45,60 €