Orders LinkContactUsLinkComponent
Analogue stopwatches, split-second addition timer
Analogue stopwatches, split-second addition timer
Analog stopwatch, ⅒ seconds, 15 minutes, lever and seven jewels, 130 grams
Artikelnr. 609-1132
Anbieter:   HANHART
Analogue stopwatches, split-second addition timer
Artikelnr. 609-1132
Anbieter:  HANHART
Analogue stopwatches, split-second addition timer
Analogue stopwatches, split-second addition timer
Analog stopwatch, ⅒ seconds, 15 minutes, lever and seven jewels, 130 grams
Preis
702,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Addition timer for time measurement with interrupt option, also drag indicator for split-time measurement.
  • Chrome plated brass casing, diamond cut
  • Lever, 7 jewels
  • Shock resistant, additional dust and water protection
Specifications
  • Description:
    Analog stopwatch, ⅒ seconds, 15 minutes, lever and seven jewels, 130 grams