Orders LinkContactUsLinkComponent
Wire baskets
Wire baskets
540×255×60 mm
Catalog # 216-7452
Supplier:   HUPFER METALLWERKE
Wire baskets
Catalog # 216-7452
Supplier:  HUPFER METALLWERKE
Wire baskets
Wire baskets
540×255×60 mm
Price
368,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Two flip handles
  • Base frame is stackable
  • Excellent thermal and chemical resistance
Wire steriliser strainer basket.
Documents
Specifications
  • W×D×H:
    540×255×60 mm