Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Cool Boxes

Cooling box ISOTHERM Extreme, 10 l, 310×225×350 mm (ext); 265×165×270 mm (int)
  216-6985
Cool Boxes
  216-6985
Cool Boxes
Cool Boxes

Cooling box ISOTHERM Extreme, 10 l, 310×225×350 mm (ext); 265×165×270 mm (int)

 
49,10 €
   
1 * 1 ST
 

Blue-white PP. Sturdy, robust design, ideal for storing and transporting temperature-sensitive materials and samples.

  • Box casing and lid fully insulated with CFC-free PU foam
  • A rubber seal ensures the boxes are airtight, a lid with non breakable handle can be securely locked
  • Excellent cooling performance (depending on the model up to 24 hours cooling)
 
  • Capacity:
    10 l
  • Description:
    Cooling box ISOTHERM Extreme
  • Dimensions:
    310×225×350 mm (ext); 265×165×270 mm (int)
  • Seq:
    01