Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Bins

Storage Bins

Blue, 205×335×155 mm
Catalog # 216-3738
Supplier:   hünersdorff GmbH
Storage Bins
Catalog # 216-3738
Supplier:  hünersdorff GmbH
Storage Bins
Storage Bins

Blue, 205×335×155 mm

Price
11,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

These storage bins are particularly suitable for a wide range of applications in industry and craft.

  • Stable
  • Professional quality
  • Resistance to grease, resins, alkalis, oils and acids
Specifications
  • Colour:
    Blue
  • Seq:
    07
  • W×D×H:
    205×335×155 mm