ContactUsLinkComponent
Container multipurpose, 500ml, with snap-on lid, length 103mm, breadth 103mm, height 64mm, container high density polyethylene, lid low density polyethylene 1 * 1 items
Catalog # 216-3412
Supplier:  hünersdorff GmbH
Container multipurpose, 500ml, with snap-on lid, length 103mm, breadth 103mm, height 64mm, container high density polyethylene, lid low density polyethylene 1 * 1 items
Catalog # 216-3412
Supplier:  hünersdorff GmbH
Frequently Bought Together