Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Bottles, narrow neck, rectangular, LDPE, with thread

1000 ml
Catalog # 215-8334
Supplier:   MUELLER KREMPEL
Bottles, narrow neck, rectangular, LDPE, with thread
Catalog # 215-8334
Supplier:  MUELLER KREMPEL
Bottles, narrow neck, rectangular, LDPE, with thread
Bottles, narrow neck, rectangular, LDPE, with thread

1000 ml

Price
3,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

LDPE, translucent, without screw cap. Ideal for storing and shipping liquids. Graduated.

  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Side ribbing for better grip

Ordering information: The bottles are supplied without caps. For suitable caps for 125/500 and 1000 ml please see 215-9226 (for 250 ml botlles see MUEL14.063.01).

Specifications
  • Capacity:
    1000 ml