Order Entry
Austria
Orders LinkContactUsLinkComponent
Wire baskets

Wire baskets

200×300×200 mm
Catalog # 215-7422
Supplier:   HUPFER METALLWERKE
Wire baskets
Catalog # 215-7422
Supplier:  HUPFER METALLWERKE
Wire baskets
Wire baskets

200×300×200 mm

Price
478,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel.

  • Spot welded mesh 8×8×1 mm
  • Electrolytically polished
  • Reinforced edges

These baskets can be used for transport, storage, cleaning and sterilisation of laboratory utensils.

Specifications
  • Seq:
    08
  • W×D×H:
    200×300×200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.