Wire baskets
200×200×200 mm
215-7420
Stainless steel.
- Spot welded mesh 8×8×1 mm
- Electrolytically polished
- Reinforced edges
These baskets can be used for transport, storage, cleaning and sterilisation of laboratory utensils.
- Seq:07
- W×D×H:200×200×200 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



